Buisspiraalkoelers met NS hals, Quickfit®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CX6/22/SC CX6/33/SC CX6/44/SC
201-8305EA 308 EUR
201-8305 201-8306 BIBBCX6/44/SC
Buisspiraalkoelers met NS hals, Quickfit®
Condensors
PYREX®, borosilicaatglas.
  • High chemical and temperature resistance
  • Both cone and sockets are the same size
Ideal for reflux work, with large cooling surface and an outer water jacket, which avoids vapour moving around the coil.
Certificaten: ISO 1773, BS 6352