Allihn en Liebig koelers met NS hals, Quickfit®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CX7/02/SC CX7/03/SC CX7/04/SC CX7/06/SC CX7/05/SC CX7/08/SC
201-8250EA 187 EUR
201-8250 201-8307 201-8308 201-8310 201-8309 201-8311
Allihn en Liebig koelers met NS hals, Quickfit®
Condensors
Condensers acc. to Allihn, PYREX® borosilicate glass, for condensing vapours.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Ideal for reflux applications. With NS cone. Inner bulb sections provide increased cooling surface. With detachable plastic screwthread connectors for easy and safe attachment of flexible tubing.