Multispiraalkoelers met NS hals
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C13/55 C13/55/2
BIBBC13/55EA 1250 EUR
BIBBC13/55 BIBBC13/55/2
Multispiraalkoelers met NS hals
Condensors
PYREX®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With ground joints. Features a double coil design with a common water inlet. Suitable for pilot plant work(Condenser, multi coil) or two independent coolings coils(Condenser, twin-coil). Connectors provide easy and safe attachment of flexible tubing.