Verbindingsstukken voor slangen, X-vorm
Leverancier: KARTELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
71410 71411 71412 71413 71414 71415 71416
229-0742EA 21.1 EUR
229-0742 229-0743 229-0744 229-0745 229-0746 229-0747 229-0748
Verbindingsstukken voor slangen, X-vorm
Fittingen en connectoren
PP, translucent.
  • Good mechanical strength and chemical resistance
  • Very good thermal resistance
  • Autoclavable
  • Suitable for foodstuff
Dimensions are shown as both nominal and exact as elasticity of tubing will influence the choice of size. Use our tubing selector 229-0111 to determine "grip". This will then give equivalent crest dimension for the particular connector concerned.
They meet with the Food and Drug regulations.90/128CEE 92/39CEE and FDA 21 CFR 177.1520.
Suitable for foodstuff.
Certificaten: Zij voldoen aan de Food and Drug regelgeving. 90/128 CEE, 92/39 CEE en FDA 21 CFR 177.1520.