Slangverbiningen, rechte vorm
Leverancier: PIEPER FILTER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PP-S24S24 PP-S32S32 PP-S48S48F1 PP-S64S64R1
229-0506EA 37.6 EUR
229-0506 229-0507 229-4125 229-4126
Slangverbiningen, rechte vorm
Fittingen en connectoren
PP, transparent.
  • Very good thermal resistance
  • Very good chemical resistance
  • Robust, durable
Perfect for conneting tubing with small inner diameters