Adapter, verbinding/kraan/buis, Quickfit®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MF18/2/SC MF18/44/SC MF18/5/SC MF17/1/SC MF18/33/SC MF18/3/SC MF17/2/SC MF17/3/SC
201-0648EA 44.3 EUR
201-0648 201-8162 201-3063 201-0646 201-0659 201-0649 BIBBMF17/2/SC BIBBMF17/3/SC
Adapter, verbinding/kraan/buis, Quickfit®
Adapters Adapters, Slijpstukken met slangaansluitingen
PYREX®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With side arm for the attachment of rubber tubing etc. Detachable plastic screw thread connector allows flexible tubing to be fitted easily and safely.