Adapter met verbindingstube, recht, Quickfit®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MF10/1B/SC MF10/2B/SC MF10/3B/SC MF10/4B/SC MF10/5B/SC MF13/2/SC MF13/3/SC MF13/4/SC
201-0567EA 35.4 EUR
201-0567 201-0595 201-0568 201-0596 201-0645 201-3012 BIBBMF13/3/SC BIBBMF13/4/SC
Adapter met verbindingstube, recht, Quickfit®
Adapters Adapters, Slijpstukken met slangaansluitingen
PYREX®, borosilicate glass, clear.
  • Very good chemical resistance, high temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Screw thread connector accepts rubber hose with a bore of 9 mm
With straight or right angled connection for the attachment of rubber tubings etc. Detachable plastic screw thread connectors allow flexible tubing to be fitted easily and safely.