Monsternamepotjes met snap cap, Corning® Gosselin™
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PC1000-04 PC1000-11 PC1000-13 PC400-02 PC400-05 PC400-09 PC1000-03 PC1000-09 PC400-06 PC200A-04 PC200A-07 PC200A-03 PC200A-05 PC200A-02
216-4278EA 253 EUR
216-4278 216-0417 216-4277 215-0793 215-0794 216-2083 216-2081 GOSSPC1000-09 216-2082 215-0791 GOSSPC200A-07 GOSSPC200A-03 GOSSPC200A-05 GOSSPC200A-02
Monsternamepotjes met snap cap, Corning® Gosselin™
Monsternamepotjes Containers voor algemeen gebruik
These containers are made of transparent or white PP.
  • Lightweight and break-resistant
  • Assembled or non-assembled
  • Molded graduations
Ideal for sampling, storage and transport of liquid and solid samples.