Telkamers, Neubauer verbeterd
Leverancier: LABOR OPTIK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1106000 1120000 1110000 1130000
631-1111EA 56.8 EUR
631-1111 631-1121 631-1112 631-1113
Telkamers, Neubauer verbeterd
Object- en dekglaasjes Telkamers
Voor het tellen van leukocyten, trombocyten en erytrocyten. Alle groepsvierkanten hebben driedubbele grenslijnen.
  • Four large squares in the corners are divided into 16 squares each (leucocyte counting)
  • Large square in the middle divided in 25 squares, each of which is further divided into 16 small squares
  • The middle line determines whether cells in the border area are to be included in one square or the other
Telrooster: 9
Grote vierkanten: 1 mm² elk
Grote vierkanten: 1 mm² elk