Telkamers, Bürker
Leverancier: LABOR OPTIK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1900000 1910000 1920000 1930000
631-1141EA 61.4 EUR
631-1141 631-1122 631-1123 631-1124
Telkamers, Bürker
Object- en dekglaasjes Telkamers
Counting chambers according to Bürker's ruling.
  • Large squares (1 mm²) divided into 16 group squares by double lines (0,05 mm apart)
  • Double lines form small squares of an area of 0,0025 mm²
  • Nine counting grids
Suitable for counting leucocytes and erythrocytes.
Telrooster: 9