Smeltkroezen
Leverancier: USBECK Laborgeräte
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5620 5621 5622 5623 5624
459-1260EA 11.1 EUR
459-1260 459-1261 459-0252 459-1262 459-0174
Smeltkroezen
Kroezen
These crucibles are made from iron.
  • Recommended maximum temperature 1100 °C
  • Can be used up to 1500 °C despite a risk of shortening the life time
Area of use: Only on open burner flame.
Bestelinformatie: Lids are available separately.