Smeltkroezen, Vitreosil®
Leverancier: TECHNICAL QUARTZ SOLUTIONS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
73081 73082 73083
216-4496EA 89.4 EUR
216-4496 216-4497 216-4498
Smeltkroezen, Vitreosil®
Kroezen
These crucibles are made from fused quartz, translucent, glazed.
  • Max. temperature: 1050 °C
Leveringsinformatie: Supplied without lid.