Cuvettes with Magnetic Rods for Absorption or Fluorescence Measurements
Leverancier: HELLMA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
109000F-10-40 109000-10-40 119-10-40 109004-10-40 109004F-10-40 119004-10-40 119F-10-40 119004F-10-40
HELL109000F-10-40EA 559 EUR
HELL109000F-10-40 634-9561 634-9562 HELL109004-10-40 HELL109004F-10-40 HELL119004-10-40 HELL119F-10-40 HELL119004F-10-40
Cuvettes with Magnetic Rods for Absorption or Fluorescence Measurements
Cuvetten Cuvetten voor spectrofotometers
Hellma® High precision quartz glass macro cells for absorption or fluorescence measurements in the UV/Vis range, 4 and 10 mm path length, with magnetic rod and lid.
  • Cells made from special optical glass
  • Wavelength range from 200 to 2500 nm
  • Path length: 4 to 10 mm
  • Magnetic stirrer rod
  • With lid
  • Volume: 1500 to 3500 µl
Hellma® precision cells are 100% manufactured in Germany and are suitable for all common spectrophotometers.
Highest precision: Hellma® precision cells are characterized by the highest precision in path length accuracy and parallelism.