Macrocellen voor fluorescentiemetingen
Leverancier: HELLMA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
101-20-40 101-10-40 101-10-K-40
634-0848EA 623 EUR
634-0848 634-0508 634-0966
Macrocellen voor fluorescentiemetingen
Cuvetten Cuvetten voor spectrofotometers
Hellma® high precision quartz glass macro cells for fluorescence measurements in the UV/Vis range, 10×10 to 20 mm path length, with lid.
  • Cells made of high-performance quartz glass
  • Wavelength range from 200 to 2500 nm
  • With PTFE lid
  • Volume: 3500 to 7000 μl
Hellma® precision cells are 100% manufactured in Germany and are suitable for all common spectrophotometers.
Highest precision: Hellma® precision cells are characterised by the highest precision in path length accuracy and parallelism. This guarantees continuously reproducible measurement results.