Droogbuizen
Leverancier: KARTELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
446 447 448
468-1210EA 6.8 EUR
468-1210 468-1211 468-1212
Droogbuizen
Buisjes Droogbuizen
PE.
  • Both ends have tapered, push-on connectors
Suitable for drying gases.