Collageen I celcultuurschalen, Corning® BioCoat™
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
354401 354450 354456 354551 356401 356450
734-0109EA 120.34 EUR
734-0109 734-0136 734-0141 734-0202 734-0275 734-0280
Collageen I celcultuurschalen, Corning® BioCoat™
Cultuurschalen Celcultuurschalen
PS, coating met rattenstaartcollageen type I, steriel, pyrogeenvrij.
  • Manufactured in a highly controlled environment and rigorously tested to assure product consistency and performance
  • Stable at room temperature when stored in dry conditions
Collagen I, found in most tissues and organs, is most plentiful in dermis, tendon and bone. It is an integral part of the framework that holds cells and tissues together and has been recognised as a useful matrix for improving cell culture. Used in vitro, collagen can exert effects on the adherence, morphology, growth, migration and differentiation of a variety of cell types.