Schalen, rond, lage vorm
Leverancier: NICKEL ELECTRO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
502 503 504 505 506 508 541 543 544 545 546 548 541A 543A 544A 545A 546A 548A
216-0403EA 47.4 EUR
216-0403 NICK503 NICK504 216-0404 NICK506 NICK508 216-0405 216-0406 NICK544 216-0408 NICK546 NICK548 NICK541A NICK543A 216-0407 NICK545A NICK546A NICK548A
Schalen, rond, lage vorm
Reservoirs/Schalen/Potten Uitdampschalen
Pure nickel (99,6%).
  • Excellent corrosion and temperature resistance
  • Lids are optionally available with and without knobs
  • Smooth surfaces for fast, easy cleaning
Round, with flat bottom. Multipurpose dish ideal for a wide variety of uses.