VWR®, Dishes, Evaporating, ondiep
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
216-0173EA 27 EUR
216-0173 216-0174 216-0175 216-0176 216-0177 216-0178
VWR®, Dishes, Evaporating, ondiep
Reservoirs/Schalen/Potten Uitdampschalen
PTFE, met tuit.
  • Excellent chemical resistance
  • Temperature resistant from –200 to +280 °C
  • Outer diameter with a tolerance of +/- 1mm
  • Exceptionally smooth inner walls to prevent contamination
Dikwandig, conisch. Robuust en onbreekbaar.