Evaporating dishes with snap cap
Leverancier: VITLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
103397 103297
216-1292EA 72.3 EUR
216-1292 216-1291
Evaporating dishes with snap cap
Reservoirs/Schalen/Potten Uitdampschalen
PFA, with PE lid, translucent.
  • Uitzonderlijke chemische bestendigheid
  • Hittebestendig tot 250 °C, autoclaveerbaar
  • Goede vervanging voor schalen uit platina
For contamination free sample preparation and efficient transportation. Due to a conical depression in the middle of the base very small amounts of a solvent are adequate to absorb the evaporated samples.