VWR®, Dishes, Evaporating, halve diepte
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
216-0537EA 33.5 EUR
216-0537 216-0554 216-0553 216-0552 216-0551 216-0550 216-0549 216-0548
VWR®, Dishes, Evaporating, halve diepte
Reservoirs/Schalen/Potten Uitdampschalen
Geglazuurd porselein.
  • Very good chemical resistance
  • Heat resistant up to +1000 °C
  • Low thermal expansion
The economical alternative for a wide range of laboratory applications. With spout and round bottom. White glaze inside.