Uitdampschalen
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1450/03D 1450/04D 1450/06D 1450/10D 1450/01D 1450/14D
216-2759EA 154 EUR
216-2759 216-2760 BIBB1450/06D 216-2761 BIBB1450/01D BIBB1450/14D
Uitdampschalen
Reservoirs/Schalen/Potten Uitdampschalen
PYREX®, borosilicate glass, with spout.
  • High resistance to chemical attack and thermal shock
  • Complies to DIN 12336
  • Quick and easy to clean
For wide range of applications. Feature flat base and curved sides.