Instrument dishes
Leverancier: P.S. KUNSTSTOFWERKE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2037290010SET 2034410010SET 2035580010SET 2035160010 2034250010 2037290010 2035580010 2046440010 2060740001 2049120001 2052770001_U 2034410010B
216-1453EA 0 EUR
216-1453 216-1454 216-1455 216-1461 216-1462 216-1463 216-1465 216-1685 216-1686 216-1688 216-1690 PSKA2034410010B
Instrument dishes
Reservoirs/Schalen/Potten Instrumentenschalen
Melamine dishes, white; PS lid, clear.
  • Stable, good break-resistance
  • Temperature resistant from −178 to +80 °C (melamine) and −40 to +55 °C (PS)
  • Smooth surfaces for easy cleaning
Multipurpose dishes for a wide variety of laboratory applications. Also suitable for use in the food industry.