Petrischalen, PYREX®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1480/02D 1480/12D 1480/06D 1480/08D 1480/10D
391-2023EA 235 EUR
391-2023 391-2027 391-2024 391-2025 391-2026
Petrischalen, PYREX®
Cultuurschalen Petrischalen
PYREX®, borosilicate glass, with lid.
  • Chemically resistant
  • Specially formed to ensure even wall thickness and consistent optical performance
  • Able to withstand repeated autoclaving