Bottle top tilt measures
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
05476010 05476019 05476028 05476022 05476037 05476001 05476007 05476013 05476004
613-6256EA 54.4 EUR
613-6256 613-6252 613-6255 613-0696 613-0697 613-6251 613-6254 613-6250 613-6253
Bottle top tilt measures
Flesdispenseropzet
DURAN®, borosilicate glass 3.3, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
This dispenser head is completed by a 1 litre bottle for repetitive dispensing of the same volume.
NS 29/32