Dispensers voor steriele handschoenen
Leverancier: S-CURVE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SD-300-C SD-300A SD-350-C
112-0536EA 620 EUR
112-0536 113-0339 SCURSD-350-C
Dispensers voor steriele handschoenen
Handschoendispensers
These safety dispensers are designed to protect prepacked sterile gloves from contamination.
  • Easy access first in/first out usage
  • Amber version to protect from ozone degradation
  • Available in free-standing or wall-mountable versions