Stekkerdozen, 6 contactpunten, Economy
Leverancier: BACHMANN ELEKTRO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
381.251K 381.249K 381.250K 381.252K 381.2501K
139-1330EA 0 EUR
139-1330 139-1372 139-1373 139-1374 139-1375
Stekkerdozen, 6 contactpunten, Economy
Stopcontacten
Trailing socket with built-in, attractive and practical cable rewind.
  • Moulded, shock-proof plug with double earthing
  • Mounting flange for quick, easy attachment
  • Sockets offset by 35°
Kleur: wit.