Kabelboxen, 4 contactpunten
Leverancier: BACHMANN ELEKTRO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
392.181 392.180
139-1397EA 0 EUR
139-1397 BACM392.180
Kabelboxen, 4 contactpunten
Stopcontacten
Cable reel with four Schuko outlets 250 V~, without lid. Stable zinc-plated stand with black PVC cable (HO5VV-F G3 1,50 mm²).
  • Design of side wheels helps dissipate heat and humidity
  • Integrated one pole thermal cut-off switch
  • Right angle plug with double earth
  • VDE compliant
Kleur: zwart.
Max. power: Fully unrolled 3500 W; cable on reel 1000 W.