Extractie-apparaat volgens Soxhlet, Quickfit®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
EX5/32 EX5/43 EX5/53 EX5/55/100 EX5/63 EX5/83 EX5/55
201-8373EA 158 EUR
201-8373 201-8374 201-8375 201-8377 201-8378 201-8380 BIBBEX5/55
Extractie-apparaat volgens Soxhlet, Quickfit®
Extractors Soxhlet-extractors en Soxhlet-type extractors
PYREX®, borosilicaatglas.
For extracting soluble components or impurities from solids via repeated soaking/siphoning with solvent.
Certificaten: Conforms to BS 2071