Gelaatsscherm
Leverancier: UVEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
9705.014 9706.514
111-0221EA 71.4 EUR
111-0221 111-0222
Gelaatsscherm
Gelaatsschermen
Lightweight face shields made from durable plastic with headrest and a 1 mm thick acetate screen.
  • Integrated forehead protection
  • Twist ratchet enables simple and reliable adjustment to fit most sizes of head
  • Flip-up mechanism allows the visor to be set in any position
  • Easy lens replacement
EN 166
Certificaten: EN 166, EN 170