Bottle top vacuum filtration systems, SFCA membrane, Rapid-Flow™, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
122-0020 122-0045 155-0020 155-0045 156-4020 156-4045 157-0020 157-0045 158-0020 158-0045 161-0020 161-0045 162-0020 162-0045 290-3320 290-3345 290-4520 290-4545 291-3320 291-3345 291-4520 291-4545 292-3320 292-4520
513-1221EA 128 EUR
513-1221 513-1222 513-1551 513-1552 513-1561 513-1562 513-1571 513-1572 513-1581 513-1582 513-1611 734-5029 513-1621 513-1622 734-5061 734-5062 734-5063 734-5064 734-5065 734-5066 734-5067 734-5068 734-5069 734-5070
Bottle top vacuum filtration systems, SFCA membrane, Rapid-Flow™, Nalgene®
Filters Flesfilters
Rapid-Flow™ disposable filter units feature the exclusive rapid-flow support column design, providing fast flow rates and high throughput.
  • Color coded according to membrane
  • Fastest flow rate and lowest clogging of comparable alternatives
  • 0.1 µm PES available for mycoplasma protection
  • 0.2 µm PES devices are stem cell and mouse embryonic assay (MEA) tested
Ideal for the filtration of cell culture media, serum, additives and buffers.
Available with Fit for Purpose Sterile documentation
Non-pyrogenic and endotoxin free - Rapid-Flow filter units meet the most rigorous testing standards for any vacuum filtration unit.