Bottle top vacuum filtration units, Rapid-Flow™ Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
295-3345 295-4545 296-3345 296-4545
734-5072EA 93.1 EUR
734-5072 734-5074 734-5076 734-5078
Bottle top vacuum filtration units, Rapid-Flow™ Nalgene®
Filters Flesfilters
Rapid-Flow™ single use bottle tops with PES membrane and PS funnel. Filters screw onto glass media bottles with 33 or 45 mm neck size. They feature the exclusive Rapid-Flow support column design, providing fast flow rates and high throughput.
  • Allow direct filtration of liquid into sterile glass media bottles
  • Graduated
  • Side-arm with quick-disconnect tubing adapter
  • Color coded according to membrane
  • Fastest flow rate and lowest clogging of comparable alternatives
  • 0.1um PES available for mycoplasma protection
  • 0.2um PES devices are stem cell and mouse