High purity qualitative filter paper
Leverancier: Ahlstrom
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
512-6088EA 8.7 EUR
512-6088 512-6087 512-6089 BINZ113373 BINZ110142 512-6084 512-6083 512-6086 512-6085 BINZ110028 BINZ110148 512-6082 512-6081 BINZ110090 BINZ110136 BINZ110130 BINZ110138 BINZ110137 BINZ110139 BINZ113358 BINZ110120 BINZ110126 BINZ110129 512-6158 512-6157 512-6159 512-6154 512-6153 512-6156 512-6155 BINZ110070 BINZ110076 512-6165 512-6164 512-6166 BINZ113450 BINZ113455 BINZ113453 BINZ113452 BINZ113451 512-6161 512-6160 512-6163 512-6162 512-6059 512-6058 512-6055 512-6054 512-6178 512-6057 512-6056 BINZ110051 BINZ110050 BINZ113447 BINZ113445 BINZ113444 BINZ110053 BINZ113443 BINZ110173 BINZ113287 BINZ113320 BINZ110054 512-6051 512-6050 512-6053 512-6052 512-6069 512-6066 512-6186 512-6065 512-6068 512-6067 BINZ110046 BINZ110045 BINZ113435 BINZ110048 BINZ110047 512-6062 512-6183 512-6061 512-6182 512-6185 512-6064 512-6063 BINZ110049 512-6181 512-6060 512-6180 512-6077 512-6076 512-6079 512-6078 BINZ113386 512-6073 512-6072 512-6075 512-6074 512-6190 512-6070
High purity qualitative filter paper
Filters Filterpapier Kwalitatief filterpapier
Ahlstrom-Munksjö’s qualitative grades of filter paper are recommended for use in analytical methods which determine or identify particulate constituents of a mixture irrespective of the amount present. Qualitative filter papers are often used in routine separation work that still requires high purity and consistent performance. These filter papers covers a wide range of laboratory applications, as liquids clarification, qualitative analytical separations for precipitates and buffers filtration, and are also used for soil analysis and in food and beverage testing. Wet-strengthened grades contain a reinforcement agent that increases their resistance to rupture when wet, such as in vacuum filtration, and their resistance when filtering acidic solutions.
Grade 5 and V5: Very fast filtering, open fiber matrix. For coarse and voluminous precipitates such as iron- aluminum- and chromium hydroxide; Si- determination in steel and pig iron analysis.
Grade 3: Fast filtering, open fiber matrix. For coarse and voluminous precipitates such as iron- aluminum- and chromium hydroxide; Si- determination in steel and pig iron analysis.
Grade V150: Medium fast filtering, open to medium fiber matrix. For qualitative tasks, coarser precipitates such as lead-, iron- and silver sulfide; alkali carbonates; calcium oxalate.
Grade 1F and V1F: Medium to slowly filtering, medium tight fiber matrix. For fast filtration of fine precipitates such as calcium oxalate, magnesium ammonium phosphate, coarser forms of barium sulphate.
Grade Munktell no. 1 and 292: Medium to slowly filtering, medium tight fiber matrix. For fast filtration of fine precipitates such as calcium oxalate, magnesium ammonium phosphate, coarser forms of barium sulfate.
Grade Munktell no. 2 and 292a: Slowly filtering, tight fiber matrix. For filtration of fine precipitates such as lead dioxide, calcium fluoride, nickel sulfide, zinc sulfide.
Grade 106: Slowly filtering for fine precipitates such as lead dioxide, calcium fluoride, nickel sulphide and zinc sulphide.
Grade 120H and V120H: Very slowly filtering, tight fiber matrix. For fine precipitates such as barium sulphate, metastannic acid and cuprous oxide.