Membraandispenser voor microbiologische filtratie, Microsart® e.motion
Leverancier: Sartorius
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
16713-BO 16713-PS
SART16713-BOEA 2080 EUR
SART16713-BO SART16713-PS
Membraandispenser voor microbiologische filtratie, Microsart® e.motion
Membraan dispensers
The dispenser fully automatically releases membrane filters from their sterile packaging. You can choose one of the following modes: Push button, foot switch or sensor mode.
  • Compact design, easy to clean
  • Easy insertion of the filter band
  • Low weight
  • Battery-operable Microsart® e.motion dispenser model operates without a cable
A window in the dispenser maintains the visibility of the information on the membrane filter band, such as lot number, membrane filter type and the quantity of filters. This ensures better scheduling of tests and easy traceability.
W×D×H: 172×233×230 mm
Weight: 2,4 kg (power supply only model), 2,6 kg (integrated battery model)
Membranen Microsart® e.motion: Cellulosenitraat, steriel
De membraanfilters zijn steriel verpakt, zonder beschermend papier op elke filter, in een speciaal ontworpen afzonderlijke verpakking in een harmonicavorm.
Certificaten: CE mark. EMC Directive EN 61326-1. EN 61010