Erlenmeyers, smalle of brede hals, dikwandig
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4980-2L 4980-4L 4980-6L 5100-2L 4980-25 4980-50 4980-125 4980-250 4980-500 5100-125 5100-250 5100-500 5100-1L 4980-10 4980-1L
214-0099EA 46.3 EUR
214-0099 214-0100 214-0103 214-0107 214-0097 214-0101 214-0095 214-0098 214-0102 214-0104 214-0106 214-0108 214-0105 214-0094 214-0096
Erlenmeyers, smalle of brede hals, dikwandig
Kolven Erlenmeyers
PYREX®, borosilicate glass. Heavy Duty.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Considerably more resistant to mechanical stresses than standard Erlenmeyer flasks. Designed with thick walls and an extra wide flared rim. This provides better resistance to breakage or chipping during cleaning and other uses. Extremely flat, heavy base. They are ideal for use on magnetic stirrers and hotplates. Very durable. Graduated for approximate reading of the contents, and have a white labelling area.