Fioles fourneau graduées cylindro-coniques en verre Pyrex
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1190/04M 1190/06M 1190/02M
214-0092EA 25.5 EUR
214-0092 214-0093 BIBB1190/02M
Fioles fourneau graduées cylindro-coniques en verre Pyrex
Kolven Cultuurflessen
PYREX®, borosilicate glass.
  • Very good chemical resistance
  • Good thermal resistance
  • With labelling area
Ideal for fermentation and culture applications.