Filterflessen, voor filtratie-eenheden, Nalgene®
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
455-0150 455-0250 455-0500 455-1000
734-5085EA 136 EUR
734-5085 734-5086 734-5087 734-5088
Filterflessen, voor filtratie-eenheden, Nalgene®
Kolven Filterkolven
Save time and minimize contamination with gamma-sterilized Thermo Scientific™ Nalgene™ Rapid-Flow™ Filter Storage Bottles made from virgin PS. Use with Nalgene Bottle-Top filters or as replacement/extra receptacles for filter units. Excellent as single-use storage containers.
  • Headspace allows for extra serum or other additives to be added to the filtrate
  • Leakproof screw caps to eliminate pH shift in bottle for longer life of stored media
  • Sterilised by gamma radiation and individually bagged for a 5-year sterile shelf life
  • Non-pyrogenic and non-cytotoxic
Cap is made from virgin HDPE.