Distilleerkolven, ASTM
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1290/04M 1290/06M
511-0747EA 254 EUR
511-0747 BIBB1290/06M
Distilleerkolven, ASTM
Kolven Destilleerkolven
PYREX®, borosilicate glass.
  • High resistance to thermal shock
  • Excellent chemical durability for contamination-free performance
For the distillation of petroleum products. Used with thermometers calibrated for 76 mm immersion. Side arm inclined at 75° to the vertical axis.
Side arm 0 - Ø/L: 7×100 mm (125 ml), 6×100 mm (250 ml)
Certificaten: Complies with ASTM E133