Afzuigflessen erlenmeyervorm
Leverancier: NEUBERT VOLUME GLASSWAERE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
4040-14-2000 0403-10-18180 4040-14-1000
511-0005EA 482 EUR
511-0005 511-0003 511-0004
Afzuigflessen erlenmeyervorm
Kolven Filterkolven
DURAN®, borosilicate glass, plastic coated.
  • For vacuum use
  • Does not comply with recent German safety regulations
  • Olive hose connection 11 mm Ø
The coating provides scratch, leak and splinter protection.