Erlenmeyer flessen met NS hals
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FE100/4 FE25/1 FE25/2 FE50/1 FE50/2 FE50/3 FE100/2 FE100/3 FE150/2 FE250/2 FE250/5 FE500/3 FE1L/3 FE150/3 FE150/4 FE100/1 FE250/3 FE500/4 FE250/4 FE1L/4 FE500/2 FE2L/4
201-8092EA 70.7 EUR
201-8092 201-8416 201-8417 201-8423 201-8424 201-8425 201-8408 201-8409 201-8410 201-8418 201-8421 201-8427 201-8413 201-8411 201-8412 212-0586 201-8419 201-8428 201-0256 BIBBFE1L/4 BIBBFE500/2 BIBBFE2L/4
Erlenmeyer flessen met NS hals
Kolven Erlenmeyers
PYREX® borosilicate glass, clear. With matt white labelling area.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
ISO 4797
Certificaten: ISO 4797