Flessen, Büchner met NS hals, Quickfit®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FB250/3 FB250/3SC FB500/3SC FB1L/3SC FB1L/3 FB500/3 FB1L/4 FB250/4 FB250/4SC FB100/2SC FB100/3SC FB1L/4SC FB500/4
511-0130EA 137 EUR
511-0130 511-0235 511-0238 511-0233 511-0232 511-0131 BIBBFB1L/4 BIBBFB250/4 BIBBFB250/4SC BIBBFB100/2SC BIBBFB100/3SC BIBBFB1L/4SC BIBBFB500/4
Flessen, Büchner met NS hals, Quickfit®
Kolven Filterkolven
PYREX®, borosilicaatglas.
  • Heavy wall and base construction for vacuum work
  • Side arm features a PTFE screwthread connector for the safe attachment of 9 mm bore vacuum tubing
Büchner flasks with precision-ground Quickfit® socket.
ISO 1773, BS 6352
Certificaten: ISO 1773, BS 6352