Rondbodemflessen, korte NS hals, Quickfit®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FR2L/4S FR50/3S FR5L/4S FR100/4S FR1L/3S FR1L/4S FR25/1S FR25/2S FR250/3S FR250/4S FR5/1S FR50/1S FR50/2S FR500/3S FR500/4S FR10/1S FR100/1S FR100/2S FR100/3S FR250/1S FR2L/3S FR5L/5US FR25/3S FR100/5S FR150/2S FR150/3S FR150/4S FR250/2S FR250/5S FR500/2S FR500/5S FR1L/5S FR10/0S FR150/5S FR1L/2S FR5/0S FR3L/5S FR2L/5S FR3L/3S
201-8515EA 63.3 EUR
201-8515 201-8533 201-8546 201-8475 201-8484 201-8491 201-8497 201-8498 201-8503 201-8509 201-8529 201-8530 201-8531 201-8536 201-8542 201-8467 201-8468 201-8469 201-8471 212-0608 201-8512 201-8547 201-8499 201-8476 201-8479 201-8481 201-8482 201-8500 201-8510 201-8534 201-8544 201-8492 201-8466 212-0606 BIBBFR1L/2S BIBBFR5/0S BIBBFR3L/5S BIBBFR2L/5S BIBBFR3L/3S
Rondbodemflessen, korte NS hals, Quickfit®
Kolven Rondbodemkolven
PYREX®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With white, matt labelling area.
Certificaten: ISO 1773, ISO 4797.