Rondbodemflessen met twee NS halzen, Quickfit®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FR250/3S/2A FR500/3S/2A FR25/1S/1A FR50/1S/1A FR100/3S/1A FR100/3S/2A S/3L5086 FR250/3S/1A FR1L/3S/2A FR1L/3S/1A FR500/3S/1A
201-2360EA 62.3 EUR
201-2360 201-8541 201-0609 201-0564 201-0563 201-0608 201-0492 201-0713 BIBBFR1L/3S/2A BIBBFR1L/3S/1A BIBBFR500/3S/1A
Rondbodemflessen met twee NS halzen, Quickfit®
Kolven Rondbodemkolven
PYREX®, borosilicate glass, with two necks, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
One vertical centre neck and one angled side neck. With matt white labelling area.
Certificaten: ISO 1773, BS 6352