Gasmonsternamebuizen
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
05680058 05680070 05680043 05680052
201-9113EA 106 EUR
201-9113 201-9115 201-9111 LENZ05680052
Gasmonsternamebuizen
Buisjes Gasmonsterbuizen
Duran® clear borosilicate glass.
  • Excellent thermal and chemical resistance
  • Connection on one side with capillary tube (internal Ø: 2,7 mm, external Ø: 8,0 mm) and on the other side with tube connector (external Ø: 8,0 mm)
  • With white labelling area
Gas collection tubes for removing, collecting and transporting gas samples. Both sides with a 3 NS/14 standard ground one-way stop cock.
Certificaten: DIN 12473-1