Rondbodemfles, smalle hals met verwijde rand
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1100/32D 1100/35D 1100/36D 1100/02M 1100/04M 1100/12D 1100/18D 1100/24D 1100/28D 1100/38D
214-0020EA 130 EUR
214-0020 214-0021 214-0022 214-0360 214-0015 214-0016 214-0017 214-0018 214-0019 214-0023
Rondbodemfles, smalle hals met verwijde rand
Kolven Rondbodemkolven
PYREX®, borosilicate glass.
  • High resistance to thermal shock
  • Excellent chemical durability for contamination-free performance
Narrow neck flasks with round bottom and white labelling area.
*This size is not included in the ISO range.
Certificaten: ISO 1773