Maatkolven, klasse B
Leverancier: VITLAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
676941 671941 672941 673941 674941 675941 677941
612-3786EA 21.7 EUR
612-3786 VITL671941 VITL672941 VITL673941 VITL674941 VITL675941 VITL677941
Maatkolven, klasse B
Kolven Maatkolven
PP, with PP stoppers.
Highly transparent in accordance with DIN 12664 (glass) with blue ring mark and lettering. Individually adjusted mark to contain (TC, In).