Maatkolven, klasse A, bruine maatverdeling
Leverancier: HIRSCHMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2822166 2822153 2822161 2822165 2822170 2822175 2822178 2822180 2822181 2822185 2822186 2822190 2822193 2822194 2822152 2822160 2822171 2822192 2822197 2822198
HIRS2822166EA 20.9 EUR
HIRS2822166 612-1575 612-1576 612-1577 612-1578 612-1579 612-1580 612-1581 612-1582 612-1583 612-1584 612-1585 612-1586 612-1587 HIRS2822152 HIRS2822160 HIRS2822171 612-5739 612-5740 612-5742
Maatkolven, klasse A, bruine maatverdeling
Kolven Maatkolven
DURAN®, borosilicate glass, with PE stopper, conformity certified (DE-M marking).
  • Amber stain markings
  • Certificate of conformity with dated batch identification available via website: (www.hirschmannlab.de)
*Wide neck
Certificaten: DIN EN ISO 1042
Order Now
Specificaties
Environmentally Preferable