Maatkolven, klasse A, witte graduatie
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3151/04 3151/06 3151/08 3151/10 3151/12 3151/14 3151/16 3151/18 3151/20 3151/22 3151/24 3151/26 3151/01 3151/02
612-3233EA 67.7 EUR
612-3233 612-3231 612-2902 612-3238 612-3236 612-3222 612-3215 612-3213 612-3198 612-3228 612-3226 612-3224 612-2900 612-2901
Maatkolven, klasse A, witte graduatie
Kolven Maatkolven
PYREX®, borosilicate glass, with PE stopper.
  • Calibrated to the tolerances required by the United States Pharmacopeia (USP), which are tighter than those of ISO/DIN standards
  • Each flask is laser etched with a unique individual serial number, batch code and date of manufacture - Batch certificates are downloadable from website: www.scilabware.com
  • Calibrated to contain (TC, In)
  • White enamel inscriptions
Certificaten: BS EN ISO 1042, DIN 12664