Volumetric flasks, class A, blue markings
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FRD184 FRD164 FRD166 FRD167 FRD168 FRD174 FRD176 FRD178 FRD180 FRD182 FRD170 FRD172
612-0171EA 328 EUR
612-0171 612-0442 612-0443 612-0444 612-3988 612-0446 612-0447 612-0448 612-0449 612-0450 612-0451 612-3667
Volumetric flasks, class A, blue markings
Kolven Maatkolven
MBL®, borosilicate glass, with PE stopper.
  • Marks and inscriptions in high contrast blue enamel, conformity 'H' mark
  • Each flask is laser etched with a unique individual serial number, batch code and date of manufacture - Batch certificates are downloadable from website: www.scilabware.com
  • Calibrated to contain (TC, In)
Certificaten: DIN 12664, BS EN ISO 1042