Maatkolven, klasse A, blauwe graduatie
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3152/04AWC 3152/06AWC 3152/08AWC 3152/10AWC 3152/12AWC 3152/14AWC 3152/16AWC 3152/18AWC 3152/20AWC 3152/22AWC
612-3203EA 292 EUR
612-3203 612-3204 612-3205 612-3206 612-3207 612-3208 612-3209 612-3210 612-3211 612-3212
Maatkolven, klasse A, blauwe graduatie
Kolven Maatkolven
PYREX®, borosilicate glass, amber, with polyethylene stopper.
  • Marks and inscriptions in high contrast blue enamel
  • Batch coded for traceability and supplied with a batch certificate
Certificaten: ISO 1042, DIN 12 664