Maatkolven, klasse B, Azlon®
Leverancier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
FDF018N FDF024N FDF012N FDF030N FDF036N
214-0116EA 25.2 EUR
214-0116 214-0117 214-0115 214-0118 214-0247
Maatkolven, klasse B, Azlon®
Kolven Maatkolven
PP, translucent, with PE stopper.
  • Class B tolerances with a clear easy to read calibration line
  • Feature a mixing chamber at the top of each flask's neck
  • Exposure to temperatures above 60 °C may affect accuracy
  • Cleaning with mild alkaline detergents recommended up to 60 °C
Certificaten: ISO 1042
Order Now
Specificaties
Environmentally Preferable